Order Entry
Sweden
ContactUsLinkComponent
Resorcinol
Resorcinol
Catalog # MOLE14076572-1KG
Supplier:  Molekula
CAS Number:  
undefined
Resorcinol
Catalog # MOLE14076572-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:12,08323
Boiling Pt:281 °C (1013 hPa)
Melting Pt:109…111 °C
Density:1.272 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-585-2
UN:2876
ADR:6.1,III
 :110.11 g/mol
 :GHS07
 :GHS09
CAS Number:108-46-3
Formula:C₆H₆O₂
MDL Number:MFCD00002269