Resorcinol, GR analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:108-46-3
:
Formula:C₆H₆O₂
MDL Number:MFCD00002269
Merck Index:12,08323
Boiling Pt:281 °C (1013 hPa)
Melting Pt:109…111 °C
Density:1.272 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
EINECS:203-585-2
UN:2876
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:110.11 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Melting Point:109 - 111 °C
- Molar Mass:110.11 g/mol
- Bulk Density:600 - 700 kg/m3
- Boiling Point:281 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:171 °C
- Explosion Limit:1.4 %(V)
- Density:1.28 g/cm3 (20 °C)
- pH:4 - 6 (100 g/l, H2O, 20 °C)
- Ignition Point:605 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.07593.0100_TDS_EN ProductPage_107593
Product specification
msds_107593