Resorcinol
Catalog # EHERC16811200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
Merck Index:12,08323
Boiling Pt:281 °C (1013 hPa)
Melting Pt:109…111 °C
Density:1.272 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-585-2
UN:2876
ADR:6.1,III
:110.11 g/mol
:GHS07
:GHS09
CAS Number:108-46-3
Formula:C₆H₆O₂
MDL Number:MFCD00002269