Phthalic anhydride
Catalog # EHERC16183000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:148.12 g/mol
Merck Index:13,07457
Boiling Pt:284 °C (1013 hPa)
Melting Pt:131 °C
Density:1.545 g/cm³ (20 °C)
Flash Pt:151 °C
EINECS:201-607-5
UN:2214
ADR:8,III
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:85-44-9
Formula:C₈H₄O₃
MDL Number:MFCD00005918