Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxythiophenol
3-Methoxythiophenol ≥98%
3-Methoxythiophenol ≥98%
:
Thermo Fisher Scientific
3-Methoxythiophenol ≥98%
:
Thermo Fisher Scientific
:
15570-12-4
:
:
C₇H₈OS
:
MFCD00004841
:
140.21 g/mol
:
225…227 °C
:
1.136
:
96 °C (204 °F)
:
Ambient
:
239-617-7
:
2810
:
6.1,III
:
GHS07
msds_ALFAA10648
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years