Sulphanilamide ≥99.9% (by titrimetric analysis) for diazotization titration
TCIAS0381-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White powder to crystal
- Purity (Diazo Method):min. 99.9 % (after drying)
- Drying loss:max. 0.05 %
- Ignition residue (Sulfate):max. 0.02 %
- Chloride:max. 0.002 %
:
:172.21 g/mol
:14,08925
:401 °C (1013 hPa)
:163…167 °C
:1,46 g/cm³ (20 °C)
:Ambient
:200-563-4
:GHS07
:63-74-1
:H₂NC₆H₄SO₂NH₂
:MFCD00007939