4-Aminophenol ≥99.0% for synthesis, Sigma-Aldrich®
: Merck
:123-30-8
:
:H₂NC₆H₄OH
:MFCD00007869
:109.13 g/mol
:13,00461
:284 °C (1013 hPa) (dec.)
:190 °C
:1,3 g/cm³ (20 °C)
:204-616-2
:2512
:6.1,III
:M-Clarity™ quality level = MQ200
:195 °C
:GHS08
:GHS07
:GHS09
:Ambient
- Assay (HClO₄):≥ 99.0 %
- Identity (IR):passes test