Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluorophenyl isocyanate
3-Fluorophenyl isocyanate ≥97%
3-Fluorophenyl isocyanate ≥97%
:
Thermo Fisher Scientific
3-Fluorophenyl isocyanate ≥97%
:
Thermo Fisher Scientific
:
404-71-7
:
:
C₇H₄FNO
:
MFCD00002015
:
137.11 g/mol
:
56…58 °C (14 mmHg)
:
1.207
:
41 °C (105 °F)
:
206-966-1
:
3080
:
6.1,II
:
GHS06
:
GHS08
:
GHS02
:
GHS07
:
Refrigerator
msds_ALFAL01496
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years