
Sodium salicylate ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:54-21-7
:
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440
:160.10 g/mol
Merck Index:12,08819
Melting Pt:200 °C
Density:0.32 g/cm³ (20 °C)
EINECS:200-198-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (calculated on dried substance):Min. 99.5 %
- Heavy metals (as Pb):Max. 10 ppm
- Loss on drying (110°C):Max. 0.5 %
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 0.06 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 50L Plastic bucket UN Y (ZPACK13247050)
Packaging Information
spec_VWRC28068.296 spec_VWRC28068.460 spec_VWRC28068.460 spec_VWRC28068.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together