2,4-Dichlorophenyl isothiocyanate
Glass bottle for solids, 10 g
Catalog # MAYBSB01627EA
Marketsource
Supplier: Thermo Scientific
2,4-Dichlorophenyl isothiocyanate
Catalog # MAYBSB01627EA
Supplier: Thermo Scientific
Marketsource
CAS Number: 6590-96-1
Jump to:
Product Details & Documents
Synonyms: 2,4-Dichlorophenylisothiocyanate, 1,3-Dichloro-4-isothiocyanatobenzene
- CAS Number:6590-96-1
- Formula:C₇H₃Cl₂NS
- MDL Number:MFCD00037827
- Molecular Weight:204.08 g/mol
- Boiling Pt:260 °C (1013 hPa)
- Melting Pt:58…59 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:229-527-6
- UN:2923
- ADR:8,III
- :Density: 1.41 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



