Anti- Mouse Polyclonal Antibody (Unconjugated)
Catalog # IMMRMU-006-M
Supplier: ImmunoReagents
Specifications
- Antikroppstyp:Kontroll
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Mouse
- Form:Liquid
- Koncentration:0.5 mg/ml
- Rening:Purified
- Förp.:100 µG
Specifications
About this item
Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: