Anti-IgG Rabbit Polyclonal Antibody (Biotin)
IMMRRB-003-CBIO
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Form:Liquid
- Concentration:1.5 mg/ml
- Purification:Purified
- Pack type:Glass vial
- Pk:1 mg
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: