Order Entry
Sweden
ContactUsLinkComponent
 
Anti- Llama Polyclonal Antibody (Unconjugated)
  IMMRIR2401
Anti- Llama Polyclonal Antibody (Unconjugated)
  IMMRIR2401

 

  • Antibody type:
    Control
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Llama
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    10 mg
  • Pk:
    10 mg

 

 

Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype:
Reactivity: