Order Entry
Sweden
ContactUsLinkComponent
4-Cyanobenzaldehyde for synthesis, Sigma-Aldrich®
4-Cyanobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
4-Cyanobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    100 - 103 °C
  • Molar Mass:
    131.14 g/mol
  • Boiling Point:
    133 °C (16 hPa)
  • Synonyms:
    4-Formylbenzonitrile
  • Hill Formula:
    C8H5NO
  • Chemical Formula:
    4-(OHC)C6H4CN

Specification Test Results

Related Information