3-Bromophenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89598-96-9
:
Formula:C₆H₆BBrO₂
MDL Number:MFCD00239386
:200.83 g/mol
Melting Pt:175…177 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %