6-Bromopiperonal 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15930-53-7
:
Formula:C₈H₅BrO₃
MDL Number:MFCD00022952
:229.03 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:125 to 129 °C
- GC:≥97.5 %