0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boats
Weighing Boats
Weighing boats
Weighing boats
Weighing boats
Supplier:
LUDI
Certificates
Expand image
Weighing boats
Supplier:
LUDI
About this item
Made of aluminium.
Lightweight
Good chemical resistance
Good temperature resistance