Order Entry
Sweden
ContactUsLinkComponent
Oleic acid, Supelco®
Oleic acid, Supelco®
Catalog # 05508-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Oleic acid, Supelco®
Catalog # 05508-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
Merck Index:13,06898
Boiling Pt:286 °C (1013 hPa)
Melting Pt:16.7 °C
Density:0.8946 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:180 °C
 :282.47 g/mol
Storage Temperature:Freezer
EINECS:204-007-1
CAS Number:112-80-1
Formula:C₁₈H₃₄O₂
MDL Number:MFCD00064242