Acrylamide ≥99.9%, white crystalline powder, Ultrapure for electrophoresis
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:82…92 °C
Density:1.122 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Ambient
EINECS:201-173-7
UN:2074
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Identity Test:Passes
- Purity:>99,9%
- FTIR:Conforms to Standard
- Melting Point :82 - 92 °C
- OD290 (1% aq soln) :<0,10
- Acrylic Acid Content:<0,001%
- pH:5,0 - 7,0 (1% aq soln)
- Solubility (40% aq soln):Clear, colourless solution