Acrylamide 99+% for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
Merck Index:14,129
Boiling Pt:125 °C (25 mmHg)
Melting Pt:82…85 °C
Density:1.322
Flash Pt:138 °C (280 °F)
Storage Temperature:Ambient
EINECS:201-173-7
UN:2074
ADR:6.1,III
:GHS06
:GHS08
:GHS07