

Trichloroethylene ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:79-01-6
:
Formula:ClCH=CCl₂
MDL Number:MFCD00000838
Merck Index:12,09769
Boiling Pt:86…88 °C (1.013 hPa)
Melting Pt:–84 °C
Density:1.463…1.465 g/cm³ (20 °C)
Flash Pt: Does not flash (release flammable vapour)
UN:1710
ADR:6.1,III
:131.39 g/mol
EINECS:201-167-4
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay (GC):Min. 99.50 %
- Identity (IR):complying
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.02 %
- Titrable Base:Max. 0.0003 meq/g
- Titrable Acid:Max. 0.0001 meq/g
- Free halogens:complying
- Heavy metals (as Pb):Max. 1 ppm
- APHA:Max. 10
- Refractive Index:n20/D 1.476 (lit.)