etylfenylacetat 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101-97-3
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009178
:164.2 g/mol
Merck Index:13,03872
Boiling Pt:226 °C (1013 hPa)
Melting Pt:–29 °C
Density:1,03 g/cm³
Flash Pt:102 °C
EINECS:202-993-8
:GHS07
Storage Temperature:Omgivande
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4970 to 1.4990 (20°C, 589 nm)
- GC:≥98.5 %
- Acid value:≤1 mg KOH/g