Order Entry
Sweden
ContactUsLinkComponent
Benzyl acetate
Benzyl acetate
Catalog # ROTH6014.1
Supplier:  Roth Carl
CAS Number:  
Benzyl acetate
Catalog # ROTH6014.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :150.18 g/mol
Merck Index:13,01125
Boiling Pt:206 °C (1013 hPa)
Melting Pt:–51 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:95 °C
Storage Temperature:Ambient
EINECS:205-399-7
CAS Number:140-11-4
Formula:C₉H₁₀O₂
MDL Number:MFCD00008712