4-(Indol-3-yl)butyric acid
Catalog # ROTH6258.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g
Specifications
About this item
:
:203.24 g/mol
Merck Index:13,04987
Melting Pt:121…126 °C
Density:1.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-101-5
UN:2811
ADR:6.1,III
CAS Number:133-32-4
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD00005664