Order Entry
Sweden
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Flammable
Irritant
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.5 %
  • Acidity:
    Max. 0.001 meq/g
  • Evaporation residue:
    Max. 20 ppm
  • Peroxides:
    Max. 5 ppm
  • Water:
    Max. 0.05 %
  • Transmittance (254 nm):
    Min. 60 %
  • Transmittance (280 nm):
    Min. 80 %

Specification Test Results

Frequently Bought Together