Order Entry
Sweden
ContactUsLinkComponent
 
 
 
tert-Butyl methyl ether ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
 
undefined
 
 
tert-Butyl methyl ether ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99.8 %
  • IR spectrum:
    complying
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Peroxides (as H₂O₂):
    Max. 0.0005 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 260 nm:
    Max. 0.05
  • Absorbance at 250 nm:
    Max. 0.10
  • Absorbance at 220 nm:
    Max. 0.40
  • Absorbance at 210 nm:
    Max. 1.0
  • APHA:
    Max. 10
  • Vapor Density:
    3.1 (vs air)
  • Refractive Index:
    n20/D 1.369 (lit.)

 

 

Frequently Bought Together