Order Entry
Sweden
ContactUsLinkComponent
tert-Butyl methyl ether 99+% for HPLC
Flammable
Irritant
tert-Butyl methyl ether 99+% for HPLC
Catalog # 41839.AK
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether 99+% for HPLC
Catalog # 41839.AK
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

CAS Number:1634-04-4
 :
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812
 :88.15 g/mol
Merck Index:14,6032
Boiling Pt:55…56 °C
Melting Pt:–109 °C
Density:0.742
EINECS:216-653-1
UN:2350
ADR:3,II
 :Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:–25 °C (–13 °F)
 :GHS02
 :GHS07
Storage Temperature:Ambient