tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1634-04-4
:
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812
:88.15 g/mol
Merck Index:13,06059
Boiling Pt:54…56 °C (760 mmHg)
Melting Pt:–110 °C
Density:0.7404 g/cm³
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Peroxides:≤0.005 % (as H2O2)