Isobutyl isothiocyanate
Catalog # MAYBSB01703EA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:115.2 g/mol
Boiling Pt:160…162 °C (1013 hPa)
Density:0.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-733-2
UN:2924
ADR:3,III
:GHS02
:GHS05
:GHS07
CAS Number:591-82-2
Formula:C₅H₉NS
MDL Number:MFCD00022062