Glycerine ≥99.5% ACS, Riedel-de Haën™
:56-81-5
:
:HOCH₂CH(OH)CH₂OH
:MFCD00004722
:92.09 g/mol
:13,04497
:290 °C (1.013 hPa)
:18 °C
:1.26 g/cm³ (20 °C)
:160 °C
:Ambient
:200-289-5
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
- Ignition Residue:≤0.0050%
- Identity (IR):complying
- Assay (GC):Min. 99.50 %
- Residue on ignition:Max. 0.005 %
- Water (Karl Fischer):Max. 0.5 %
- Indifference:complying
- Heavy metals (as Pb):Max. 2 ppm
- Sulfate (SO₄):Max. 0.001 %
- Halogene compounds (as Cl):Max. 0.003 %
- Acrolein and glucose:complying
- Fatty acids and ester (as butyric acid):Max. 0.05 %
- Reaction against H₂SO₄:complying
- APHA:Max. 10