Order Entry
Sweden
ContactUsLinkComponent
Formaldehyde diethyl acetal stabilised for synthesis, Sigma-Aldrich®
Flammable
Formaldehyde diethyl acetal stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Formaldehyde diethyl acetal stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    91 g/l (18 °C)
  • Melting Point:
    -66 °C
  • Molar Mass:
    104.15 g/mol
  • Boiling Point:
    88 °C (1013 hPa)
  • Vapor Pressure:
    70 hPa (20 °C)
  • Flash Point:
    -6 °C
  • Explosion Limit:
    1.5 %(V)
  • Density:
    0.83 g/cm3 (20 °C)
  • Ignition Point:
    174 °C

Specification Test Results

Related Information