

Diisopropyl ether ≥99.0% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:108-20-3
:
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
Flash Pt:–28 °C
UN:1159
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 10 ppm
EINECS:203-560-6
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.0%
- Acidity:Max. 0.0007 meq/g
- Boiling point:67 - 69 °C
- Colouration:Max. 10 APHA
- Density (20/20):0.723 - 0.728
- Evaporation residue:Max. 50 ppm
- Peroxides (as H2O2):Max. 10 ppm
- Water:Max. 0.05%
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.2 ppm
- Ba (Barium):Max. 0.2 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.2 ppm
- Co (Cobalt):Max. 0.2 ppm
- Cr (Chromium):Max. 0.2 ppm
- Cu (Copper):Max. 0.2 ppm
- Fe (Iron):Max. 0.5 ppm
- Mg (Magnesium):Max. 0.2 ppm
- Mn (Manganese):Max. 0.2 ppm
- Ni (Nickel):Max. 0.2 ppm
- Pb (Lead):Max. 0.2 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.2 ppm
- Conforms to ACS:Passes Test
- Conforms to Reag. Ph. Eur.:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
spec_VWRC24900.296 spec_VWRC24900.296 spec_VWRC24900.320 spec_VWRC24900.320 spec_VWRC24900.320
Product specification
More