

Dipropyl ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-43-3
:
Formula:C₆H₁₄O
MDL Number:MFCD00009376
:102.18 g/mol
Merck Index:12,08042
Boiling Pt:90…91 °C
Melting Pt:–122 °C
Density:0.747
Flash Pt:–28 °C (–18 °F)
Storage Temperature:Ambient
EINECS:203-869-6
UN:2384
ADR:3,II
:GHS02
:GHS07