

Dipropyl ether 99+%
Catalog # ACRO150040500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3790 to 1.3810 (20°C, 589 nm)
- GC:≥99.0 %
Specification Test Results
About this item
:
:102.18 g/mol
Merck Index:12,08042
Boiling Pt:89 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.73 g/cm³
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:203-869-6
UN:2384
ADR:3,II
:GHS07
:GHS02
CAS Number:111-43-3
Formula:C₆H₁₄O
MDL Number:MFCD00009376