
3-Nitropropanol for synthesis, Sigma-Aldrich®
Catalog # 8.14138.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:105.09 g/mol
- Boiling Point:138 - 140 °C (43 hPa)
- Refractive Index:1.445 (23 °C, 589 nm)
- Density:1.21 g/cm3 (20 °C)
Specification Test Results
About this item
:
:105.09 g/mol
Boiling Pt:240 °C (1013 hPa)
Density:1.21 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:246-716-9
UN:2810
:GHS06
CAS Number:25182-84-7
Formula:C₃H₇NO₃