2-(Cyclohexylamino)ethanesulphonic acid (CHES), Millipore®
Catalog # 239779-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:207.29 g/mol
Merck Index:13,02063
Melting Pt:320 °C
Storage Temperature:Ambient
EINECS:203-115-6
CAS Number:103-47-9
Formula:C₈H₁₇NO₃S
MDL Number:MFCD00003835