Order Entry
Sweden
ContactUsLinkComponent
Triammonium citrate 98.0-101.0%, AnalaR NORMAPUR® analytical reagent
Irritant
Triammonium citrate 98.0-101.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 103013A
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Triammonium citrate 98.0-101.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 103013A
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    98.0 - 101.0 %
  • Identification:
    Passes test
  • Reducing substances:
    Passes test
  • pH (20°C; 5 %):
    6.0 - 7.5
  • Ignition residue (SO4):
    Max. 50 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 30 ppm
  • Na (Sodium):
    Max. 40 ppm
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :243.22 g/mol
Melting Pt:185 °C
Density:1 g/cm³ (25 °C)
EINECS:222-394-5
 :GHS07
Storage Temperature:Ambient
CAS Number:3458-72-8
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406