Order Entry
Sweden
ContactUsLinkComponent
Disodium oxalate ≥99.8%, AnalaR NORMAPUR® analytical reagent
Irritant
Disodium oxalate ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Disodium oxalate ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.8 %
  • pH (20°C; 3 %):
    7.5 - 8.5
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Loss on drying (105°C):
    Max. 0.05 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 50 ppm

Specification Test Results