Adipic acid
Supplier: Molekula
Certificates
About this item
CAS Number:124-04-9
:
Formula:HOOC(CH₂)₄COOH
MDL Number:MFCD00004420
Merck Index:13,00163
Boiling Pt:337 °C (1013 hPa)
Melting Pt:151 °C
Density:1.36 g/cm³ (25 °C)
Flash Pt:196 °C (closed cup)
:146.14 g/mol
EINECS:204-673-3
:GHS07
Storage Temperature:Ambient