Behenic acid ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-85-6
:
Formula:C₂₂H₄₄O₂
MDL Number:MFCD00002807
:340.59 g/mol
Merck Index:14,01023
Boiling Pt:306°C
Melting Pt:80 °C
Density:0,8221 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-010-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:77.0 to 82.0 deg-C
- Solubility in hot Methanol:almost transparency