Order Entry
Sweden
ContactUsLinkComponent
Isopropyl myristate
Isopropyl myristate
Catalog # EHERC15391990
Supplier:  EHRENSTORFER
CAS Number:  
Isopropyl myristate
Catalog # EHERC15391990
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :270.46 g/mol
Merck Index:13,05235
Boiling Pt:320 °C (1013 hPa)
Melting Pt:0…1 °C
Density:0.852 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Refrigerator
EINECS:203-751-4
CAS Number:110-27-0
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008982