Isopropyl myristate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-27-0
:
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008982
:270.46 g/mol
Merck Index:13,05235
Boiling Pt:167 °C (9 mmHg)
Melting Pt:0 - 1 °C
Density:0.85 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:203-751-4
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4360 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥95 %
- Color scale:≤30 APHA
- Acid value:≤0.5 mg KOH/g
- Iodine value:≤1 g I2/100 g