
Ethyl hexanoate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-66-0
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00009511
:144.21 g/mol
Merck Index:13,03812
Boiling Pt:168 °C (760 mmHg)
Melting Pt:67 °C
Density:0.869 g/cm³
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:204-640-3
UN:3272
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4060 to 1.4090 (20°C, 589 nm)
- Water:≤0.3 % (K.F.)
- Density:(20°C) 0.867 to 0.871 kg/l
- GC:≥99.0 %
- Acid value:≤1 mg KOH/g