4'-Methylacetanilide for synthesis, Sigma-Aldrich®
Catalog # 8.00077.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:1.2 g/l (25 °C) (experimental)
- Melting Point:147 - 150 °C
- Molar Mass:149.19 g/mol
- Boiling Point:306 - 308 °C (1013 hPa)
- Flash Point:168 °C
Specification Test Results
About this item
:
:149.19 g/mol
Merck Index:13,00076
Boiling Pt:307 °C (1013 hPa)
Melting Pt:145…148 °C
Density:1.22 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-155-4
CAS Number:103-89-9
Formula:C₉H₁₁NO
MDL Number:MFCD00008677