
4'-Methylacetanilide 99%
:103-89-9
:
:C₉H₁₁NO
:MFCD00008677
:149.19 g/mol
:13,00076
:307 °C (760 mmHg)
:148…151 °C
:1.21 g/cm³
:168 °C
:203-155-4
:GHS07
:Ambient
- Appearance:White to pink to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:147 to 151 °C
- GC:≥98.5 %