2',6'-Dimethylacetanilide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2198-53-0
:
Formula:C₁₀H₁₃NO
MDL Number:MFCD00008675
:163.22 g/mol
Boiling Pt:339 °C (1013 hPa)
Melting Pt:179…183 °C
Storage Temperature:Ambient
EINECS:218-596-8
:GHS07