


N,N-Dimethylformamide, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:57.5 °C (closed cup)
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-61 °C
- Molar Mass:73.09 g/mol
- Boiling Point:153 °C (1013 hPa)
- Vapor Pressure:3.77 hPa (20 °C)
- Flash Point:58 °C
- Refractive Index:1.43
- Explosion Limit:2.2 - 16 %(V)
- Density:0.94 g/cm3 (20 °C)
- pH:7 (200 g/l, H2O, 20 °C)
- Ignition Point:410 °C