


N,N-Dimethylformamide ≥99.8% ACS
:68-12-2
:
:HCON(CH₃)₂
:MFCD00003284
:73.09 g/mol
:13,03269
:153 °C (760 mmHg)
:–61 °C
:0.945 g/cm³
:58 °C
:Ambient
:200-679-5
:2265
:3,III
:01-2119475605-32
:GHS07
:GHS08
:GHS02
- Appearance:Clear liquid
- Water:≤0.15 % (coulometric)
- GC:≥99.8 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.005 %
- Titrable acid:≤0.0005 meq/g
- Titrable base:≤0.003 meq/g