



2-Chloroethyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1943-83-5
:
Formula:C₃H₄ClNO
MDL Number:MFCD00002043
:105.52 g/mol
Boiling Pt:135 °C
Density:1.237
Flash Pt:56 °C (132 °F)
EINECS:217-734-4
UN:3080
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator