Order Entry
Sweden
ContactUsLinkComponent
Dibromoacetonitrile analytical standard, Supelco®
Irritant
Dibromoacetonitrile analytical standard, Supelco®
Catalog # 442552.
Supplier:  Merck
CAS Number:  
undefined
Irritant
Dibromoacetonitrile analytical standard, Supelco®
Catalog # 442552.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :198.84 g/mol
Boiling Pt:67 °C (1013 hPa)
Density:2.369 g/cm³ (20 °C)
EINECS:221-843-2
UN:3276
ADR:6.1,III
 :GHS07
CAS Number:3252-43-5
Formula:Br₂CHCN
MDL Number:MFCD00001856